Assay Details
Target Gene Details
Entrez Gene ID: | 72354 |
Gene Name: | tetratricopeptide repeat domain 4 |
Gene Aliases: |
2810002P21Rik, AF007005, L62 |
Location: |
Chr.4:106655014-106678967 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc4 | NM_001172073.1 | NP_001165544.1 | ||
| NM_028209.2 | NP_082485.1 | |||
| AK005923.1 | ||||
| AK012652.1 | BAB28383.1 | |||
| AK088684.1 | BAC40504.1 | |||
| AK133931.1 | BAE21933.1 | |||
| AK184523.1 | ||||
| BC025435.1 | AAH25435.1 | |||
| BC125257.1 |

Genomic Map