Assay Details
Target Gene Details
Entrez Gene ID: | 59002 |
Gene Name: | WD repeat containing, antisense to Trp73 |
Gene Aliases: |
2610044M17Rik, 5330425N03Rik, Dd57, Wdr8 |
Location: |
Chr.4:154142357-154168259 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wrap73 | NM_021499.2 | 3 | 355 | NP_067474.2 |
| AB034911.2 | 3 | 335 | BAA92311.1 | |
| AK011773.1 | 3 | 319 | ||
| AK051688.1 | 3 | 299 | BAC34722.1 | |
| AK156969.1 | 3 | 354 | BAE33920.1 | |
| AK160331.1 | 3 | 327 | BAE35741.1 | |
| AK172690.1 | 3 | 354 | BAE43131.1 | |
| AK182927.1 | ||||
| BY063723.1 |

Genomic Map