Assay Details
Target Gene Details
Entrez Gene ID: | 21894 |
Gene Name: | talin 1 |
Gene Aliases: |
Tln |
Location: |
Chr.4:43531513-43562583 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 52 - Exon 53 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tln1 | NM_011602.5 | NP_035732.2 | ||
| AK122420.2 | BAC65702.4 | |||
| AK147228.1 | BAE27781.1 | |||
| AK170630.1 | BAE41923.1 | |||
| AK171322.1 | BAE42391.1 | |||
| AK190044.1 | ||||
| BC018557.1 | AAH18557.1 | |||
| BC121787.1 | ||||
| BC150810.1 | ||||
| X56123.1 | CAA39588.1 |

Genomic Map