Assay Details
Target Gene Details
Entrez Gene ID: | 67830 |
Gene Name: | RER1 retention in endoplasmic reticulum 1 homolog (S. cerevisiae) |
Gene Aliases: |
1110060F11Rik, 5830454N22Rik, AU043380 |
Location: |
Chr.4:155074109-155086382 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rer1 | NM_026395.1 | NP_080671.1 | ||
| AK002550.1 | BAB22181.1 | |||
| AK003682.1 | BAB22935.1 | |||
| AK013262.1 | BAB28755.1 | |||
| AK078396.1 | BAC37253.1 | |||
| AK168192.1 | BAE40152.1 | |||
| AK178378.1 | ||||
| AK178715.1 | ||||
| AK182182.1 | ||||
| BC029189.1 | AAH29189.1 |

Genomic Map