Assay Details
Target Gene Details
Entrez Gene ID: | 50500 |
Gene Name: | tocopherol (alpha) transfer protein |
Gene Aliases: |
alpha-TTP |
Location: |
Chr.4:20008089-20030785 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttpa | NM_015767.3 | 2 | 295 | NP_056582.1 |
| AF218416.1 | 2 | 283 | AAF25956.1 | |
| AK004882.1 | 2 | 276 | BAB23640.1 | |
| AK050374.1 | 2 | 296 | BAC34218.1 | |
| BC026882.1 | 2 | 182 | AAH26882.1 | |
| BC099868.1 | 2 | 294 | AAH99868.1 | |
| BC099869.1 | 2 | 296 | AAH99869.1 | |
| BC099870.1 | 2 | 296 | AAH99870.1 | |
| BC099871.1 | 2 | 295 | AAH99871.1 |
More Information
Set Membership: |
|

Genomic Map