Assay Details
Target Gene Details
Entrez Gene ID: | 541307 |
Gene Name: | chemokine (C-C motif) ligand 26 |
Gene Aliases: |
Ccl26l |
Location: |
Chr.5:135560448-135563569 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccl26 | NM_001013412.2 | NP_001013430.2 | ||
| AY587128.1 | AAT68481.1 |
More Information
Set Membership: |
|

Genomic Map