Assay Details
Target Gene Details
Entrez Gene ID: | 12962 |
Gene Name: | crystallin, beta B3 |
Gene Aliases: |
AI852419 |
Location: |
Chr.5:113075839-113082233 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Crybb3 | NM_001159650.1 | NP_001153122.1 | ||
| NM_021352.3 | NP_067327.1 | |||
| CJ046471.1 |
More Information
Set Membership: |
|

Genomic Map