Assay Details
Target Gene Details
Entrez Gene ID: | 66898 |
Gene Name: | BAI1-associated protein 2-like 1 |
Gene Aliases: |
1300006M19Rik, AI585895, IRTKS |
Location: |
Chr.5:144264525-144358112 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Baiap2l1 | NM_025833.3 | NP_080109.1 | ||
| AK004918.2 | BAB23669.1 | |||
| AK008203.1 | ||||
| BC015459.1 | AAH15459.1 |
Target Gene Details
Entrez Gene ID: | 55950 |
Gene Name: | brain protein I3 |
Gene Aliases: |
I3 |
Location: |
Chr.5:144244437-144446757 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bri3 | AK049027.1 | BAC33516.1 |
More Information
Set Membership: |
|

Genomic Map