Assay Details
Target Gene Details
Entrez Gene ID: | 22385 |
Gene Name: | bromodomain adjacent to zinc finger domain, 1B |
Gene Aliases: |
C87820, WSTF, Wbscr9 |
Location: |
Chr.5:135187323-135246129 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Baz1b | NM_011714.2 | NP_035844.2 | ||
| AF084480.1 | AAD08676.1 | |||
| AK017894.1 | BAB30992.1 | |||
| AK036160.1 | ||||
| AK137139.1 | BAE23247.1 | |||
| AK140172.1 | BAE24264.1 | |||
| AK141305.1 | BAE24643.1 | |||
| BC141399.1 |

Genomic Map