Assay Details
Target Gene Details
Entrez Gene ID: | 27407 |
Gene Name: | ATP-binding cassette, sub-family F (GCN20), member 2 |
Gene Aliases: |
0710005O05Rik, Drr3, E430001O06 |
Location: |
Chr.5:24565341-24577467 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abcf2 | NM_001190443.1 | NP_001177372.1 | ||
| NM_013853.2 | NP_038881.1 | |||
| AK087990.1 | BAC40079.1 | |||
| AK137233.1 | BAE23280.1 | |||
| AK147937.1 | BAE28242.1 | |||
| AK151545.1 | BAE30491.1 | |||
| BC003300.1 | AAH03300.1 |
More Information
Set Membership: |
|

Genomic Map