Assay Details
Target Gene Details
Entrez Gene ID: | 27214 |
Gene Name: | DBF4 homolog (S. cerevisiae) |
Gene Aliases: |
AA545217, Ask |
Location: |
Chr.5:8396969-8422722 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dbf4 | NM_001190717.1 | 10 | 1062 | NP_001177646.1 |
| NM_013726.3 | 10 | 1065 | NP_038754.1 | |
| AJ003132.1 | 10 | 955 | CAB56847.1 | |
| AK014480.1 | 10 | 1031 | BAB29383.1 | |
| AK083650.1 | 7 | 440 | BAC38982.1 | |
| AK132403.1 | 10 | 1027 | BAE21148.1 | |
| AK163408.1 | 8 | 641 | BAE37338.1 | |
| AK165327.1 | ||||
| BC069983.1 | AAH69983.1 | |||
| BC138709.1 | 10 | 883 | ||
| BC138710.1 | 10 | 883 | ||
| BC145392.1 | 10 | 880 |
More Information
Set Membership: |
|

Genomic Map