Assay Details
Target Gene Details
Entrez Gene ID: | 12571 |
Gene Name: | cyclin-dependent kinase 6 |
Gene Aliases: |
5830411I20, AI504062, Crk2 |
Location: |
Chr.5:3343891-3531008 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdk6 | NM_009873.2 | NP_034003.1 | ||
| AF132482.1 | AAD43503.1 | |||
| AF132483.1 | AAD43504.1 | |||
| AK078973.1 | BAC37489.1 | |||
| AK154197.1 | BAE32432.1 | |||
| BC119360.1 | ||||
| BC120595.1 |
More Information
Set Membership: |
|

Genomic Map