Assay Details
Target Gene Details
Entrez Gene ID: | 231889 |
Gene Name: | BUD31 homolog (yeast) |
Gene Aliases: |
C77604, EDG-2, EDG2, G10 |
Location: |
Chr.5:145140338-145148074 on Build GRCm38 |
Assay Gene Location: | Overlaps - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bud31 | NM_001008705.1 | NP_001008705.1 | ||
| BC057025.1 | AAH57025.1 |
More Information
Set Membership: |
|

Genomic Map