Assay Details
Target Gene Details
Entrez Gene ID: | 231889 |
Gene Name: | BUD31 homolog (yeast) |
Gene Aliases: |
C77604, EDG-2, EDG2, G10 |
Location: |
Chr.5:145140338-145148074 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bud31 | NM_001008705.1 | 5 | 583 | NP_001008705.1 |
| BC057025.1 | 5 | 583 | AAH57025.1 |
Target Gene Details
Entrez Gene ID: | 71799 |
Gene Name: | pentatricopeptide repeat domain 1 |
Gene Aliases: |
1110069M14Rik |
Location: |
Chr.5:145147378-145167119 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptcd1 | NM_133735.2 | 9 | 2645 | NP_598496.3 |
| AK004421.1 | 1 | 333 | ||
| AK088764.1 | 9 | 2564 | BAC40555.1 | |
| AK143877.1 | 9 | 2603 | ||
| AK150514.1 | 9 | 2602 | BAE29626.1 | |
| AK151064.1 | 9 | 2613 | ||
| AK152077.1 | 9 | 2613 | ||
| AK153366.1 | 9 | 2613 | ||
| AK166306.1 | 1 | 470 | BAE38694.1 | |
| AK171484.1 | 9 | 2600 | BAE42485.1 | |
| AK213622.1 | 1 | 294 | ||
| AK216509.1 | 1 | 173 | ||
| AK218479.1 | ||||
| BC004766.1 | 9 | 2563 | AAH04766.2 | |
| BC083097.1 | 9 | 2604 | AAH83097.1 |
More Information
Set Membership: |
|

Genomic Map