Assay Details
Target Gene Details
Entrez Gene ID: | 12909 |
Gene Name: | calcitonin gene-related peptide-receptor component protein |
Gene Aliases: |
AL022669 |
Location: |
Chr.5:130029306-130060787 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Crcp | NM_007761.2 | NP_031787.1 | ||
| AF028242.1 | AAB84046.1 | |||
| AF118271.1 | AAD17244.1 | |||
| AK075587.1 | ||||
| BC012676.1 | AAH12676.1 |
More Information
Set Membership: |
|

Genomic Map