Assay Details
Target Gene Details
Entrez Gene ID: | 100900 |
Gene Name: | HscB iron-sulfur cluster co-chaperone homolog (E. coli) |
Gene Aliases: |
AI325508, AW049829, Hsc20 |
Location: |
Chr.5:110829070-110839802 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hscb | NM_153571.2 | NP_705799.2 | ||
| AK131961.1 | BAE20904.1 | |||
| BC027641.1 | AAH27641.1 |
More Information
Set Membership: |
|

Genomic Map