Assay Details
Target Gene Details
Entrez Gene ID: | 13667 |
Gene Name: | eukaryotic translation initiation factor 2B, subunit 4 delta |
Gene Aliases: |
Eif2b |
Location: |
Chr.5:31187558-31193139 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif2b4 | NM_001127355.1 | NP_001120827.1 | ||
| NM_001127356.1 | NP_001120828.1 | |||
| NM_010122.2 | NP_034252.2 | |||
| AK154364.1 | ||||
| AK158282.1 | BAE34444.1 | |||
| BC021884.1 | AAH21884.1 | |||
| CD768292.1 | ||||
| M98035.1 | AAA68046.1 | |||
| M98036.1 | AAA68047.1 |
More Information
Set Membership: |
|

Genomic Map