Assay Details
Target Gene Details
Entrez Gene ID: | 27979 |
Gene Name: | eukaryotic translation initiation factor 3, subunit B |
Gene Aliases: |
AL033316, AL033334, AL033369, AW208965, D5Wsu45e, EIF3-ETA, EIF3-P110, EIF3-P116, Eif3s9, PRT1 |
Location: |
Chr.5:140419305-140443358 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif3b | NM_133916.2 | NP_598677.1 | ||
| AK033722.1 | BAC28445.1 | |||
| AK089978.1 | ||||
| AK167618.1 | BAE39671.1 | |||
| AK170938.1 | BAE42128.1 | |||
| AK194329.1 | ||||
| AK203889.1 | ||||
| BC007175.1 | AAH07175.1 | |||
| BC023767.1 | AAH23767.1 | |||
| BC031704.1 | AAH31704.1 | |||
| BC051065.1 | AAH51065.1 |
Target Gene Details
Entrez Gene ID: | 102466792 |
Gene Name: | microRNA 7038 |
Gene Aliases: |
Gm27955, mmu-mir-7038 |
Location: |
Chr.5:140427339-140427409 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mir7038 | NR_106005.1 |
More Information
Set Membership: |
|

Genomic Map