Assay Details
Target Gene Details
Entrez Gene ID: | 13846 |
Gene Name: | Eph receptor B4 |
Gene Aliases: |
AI042935, Htk, MDK2, Myk1, Tyro11 |
Location: |
Chr.5:137350109-137374522 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ephb4 | NM_001159571.1 | 11 | 2337 | NP_001153043.1 |
| NM_010144.6 | 11 | 2310 | NP_034274.4 | |
| AK046884.1 | 11 | 2275 | BAC32908.1 | |
| AK049340.1 | 11 | 2311 | BAC33693.1 | |
| AK132392.1 | 11 | 2310 | BAE21142.1 | |
| BC016505.1 | 11 | 2262 | AAH16505.1 | |
| U06834.1 | 12 | 1923 | AAA18591.1 |
More Information
Set Membership: |
|

Genomic Map