Assay Details
Target Gene Details
Entrez Gene ID: | 381633 |
Gene Name: | predicted gene 1673 |
Gene Aliases: |
- |
Location: |
Chr.5:33983472-33985009 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm1673 | NM_001033458.3 | 2 | 120 | NP_001028630.1 |
| AK131594.1 | 1 | 89 | BAE20707.1 | |
| AK141724.1 | 2 | 121 | BAE24810.1 | |
| BC147444.1 | 2 | 161 | ||
| BC147468.1 | 2 | 161 |

Genomic Map