Assay Details
Target Gene Details
Entrez Gene ID: | 381633 |
Gene Name: | predicted gene 1673 |
Gene Aliases: |
- |
Location: |
Chr.5:33983472-33985009 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm1673 | NM_001033458.3 | NP_001028630.1 | ||
| AK131594.1 | BAE20707.1 | |||
| AK141724.1 | BAE24810.1 | |||
| BC147444.1 | ||||
| BC147468.1 |

Genomic Map