Assay Details
Target Gene Details
Entrez Gene ID: | 57330 |
Gene Name: | GRB10 interacting GYF protein 1 |
Gene Aliases: |
Perq1, mKIAA4110 |
Location: |
Chr.5:137510258-137527935 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gigyf1 | NM_031408.2 | 7 | 677 | NP_113596.2 |
| AY176042.1 | 7 | 693 | AAO46886.1 | |
| BY308180.1 | 4 | 299 |

Genomic Map