Assay Details
Target Gene Details
Entrez Gene ID: | 54426 |
Gene Name: | hepatocyte growth factor activator |
Gene Aliases: |
HGFA |
Location: |
Chr.5:35041514-35048436 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hgfac | NM_019447.2 | NP_062320.2 | ||
| AB009294.1 | ||||
| AF099017.1 | AAF02489.1 | |||
| AF224724.1 | AAF34712.1 | |||
| AK007987.1 | BAB25391.1 | |||
| AK075718.1 | ||||
| AK144554.1 | ||||
| AK149522.1 | BAE28936.1 | |||
| AK165433.1 | ||||
| AK169989.1 | BAE41500.1 | |||
| BC019376.1 | AAH19376.1 |
More Information
Set Membership: |
|

Genomic Map