Assay Details
Target Gene Details
Entrez Gene ID: | 14677 |
Gene Name: | guanine nucleotide binding protein (G protein), alpha inhibiting 1 |
Gene Aliases: |
AU046200, Gialpha1, Gnai-1 |
Location: |
Chr.5:18265135-18360413 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gnai1 | NM_010305.1 | 4 | 667 | NP_034435.1 |
| BC138862.1 | 4 | 547 | ||
| BC138865.1 | 4 | 547 | ||
| BY740128.1 |
More Information
Set Membership: |
|

Genomic Map