Assay Details
Target Gene Details
Entrez Gene ID: | 114674 |
Gene Name: | GTF2I repeat domain containing 2 |
Gene Aliases: |
1700012P16Rik |
Location: |
Chr.5:134183215-134218146 on Build GRCm38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gtf2ird2 | NM_053266.1 | NP_444496.1 | ||
| AK209051.1 | ||||
| AY014963.1 | AAG41674.1 | |||
| AY116023.1 | AAM48282.1 | |||
| BC132157.1 | ||||
| BC132159.1 |
More Information
Set Membership: |
|

Genomic Map