Assay Details
Target Gene Details
Entrez Gene ID: | 67980 |
Gene Name: | glucosamine-6-phosphate deaminase 2 |
Gene Aliases: |
4921523I18Rik, 4933412A11Rik, BB189630, GNPDA, Gnp2, Sb52, mKIAA4008 |
Location: |
Chr.5:69574998-69592332 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gnpda2 | NM_001038015.1 | 6 | 807 | NP_001033104.1 |
| AK014950.1 | 6 | 817 | ||
| AK015527.1 | 6 | 817 | BAB29883.1 | |
| AK017588.1 | 6 | 816 | BAB30824.1 | |
| AK051596.1 | 6 | 816 | ||
| AK141408.1 | 6 | 819 | BAE24675.1 | |
| AK166601.1 | 6 | 809 | BAE38886.1 | |
| AK168516.1 | 6 | 826 | BAE40396.1 | |
| AK171299.1 | 6 | 802 | BAE42378.1 | |
| AK220275.1 | 6 | 3369 | BAD90200.1 | |
| BC004084.1 | 6 | 807 | AAH04084.1 |
More Information
Set Membership: |
|

Genomic Map