Assay Details
Target Gene Details
Entrez Gene ID: | 66596 |
Gene Name: | general transcription factor III A |
Gene Aliases: |
2010015D03Rik, 2610111I01Rik, 5330403M05Rik |
Location: |
Chr.5:146948657-146955614 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gtf3a | NM_025652.3 | NP_079928.2 | ||
| AF391799.1 | AAL69686.1 | |||
| AK008247.2 | BAC25209.1 | |||
| AK011845.1 | ||||
| AK012797.1 | BAB28476.2 | |||
| AK017234.1 | ||||
| AK143627.1 | ||||
| BC032292.1 | AAH32292.1 |
Target Gene Details
Entrez Gene ID: | 76366 |
Gene Name: | mitochondrial translational initiation factor 3 |
Gene Aliases: |
2810012L14Rik, AI414549 |
Location: |
Chr.5:146951573-146963805 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mtif3 | NM_001256100.1 | 5 | 3864 | NP_001243029.1 |
| NM_001256101.1 | 5 | 3853 | NP_001243030.1 | |
| NM_001256102.1 | 4 | 3693 | NP_001243031.1 | |
| NM_029581.4 | 4 | 3704 | NP_083857.1 | |
| BC052439.1 | 4 | 3658 |
More Information
Set Membership: |
|

Genomic Map