Assay Details
Target Gene Details
Entrez Gene ID: | 14886 |
Gene Name: | general transcription factor II I |
Gene Aliases: |
6030441I21Rik, BAP-135, Diws1t, GtfII-I, Spin, TFII-I, WBSCR6 |
Location: |
Chr.5:134237832-134314760 on Build GRCm38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gtf2i | NM_001080746.2 | 26 | 2669 | NP_001074215.1 |
| NM_001080747.2 | 25 | 2606 | NP_001074216.1 | |
| NM_001080748.2 | 24 | 2549 | NP_001074217.1 | |
| NM_001080749.2 | 24 | 2549 | NP_001074218.1 | |
| NM_010365.4 | 25 | 2612 | NP_034495.2 | |
| AF017085.1 | 26 | 2313 | AAC53569.1 | |
| AF043219.1 | 24 | 2233 | AAC02990.1 | |
| AF043220.1 | 23 | 2170 | AAC02991.1 | |
| AK013348.1 | 3 | 134 | BAB28803.2 | |
| AK145698.1 | 16 | 1497 | ||
| AK147201.1 | 24 | 2507 | BAE27759.1 | |
| AK180983.1 | 3 | 188 | ||
| AY030290.1 | 25 | 2299 | AAK49785.1 | |
| AY030291.1 | 25 | 2305 | AAK49786.1 | |
| AY030292.1 | 24 | 2242 | AAK49787.1 | |
| AY030293.1 | 26 | 2362 | AAK49788.1 | |
| BC053044.1 | 26 | 2638 | AAH53044.1 |
More Information
Set Membership: |
|

Genomic Map