Assay Details
Target Gene Details
Entrez Gene ID: | 14886 |
Gene Name: | general transcription factor II I |
Gene Aliases: |
6030441I21Rik, BAP-135, Diws1t, GtfII-I, Spin, TFII-I, WBSCR6 |
Location: |
Chr.5:134237832-134314760 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 29 - Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gtf2i | NM_001080746.2 | NP_001074215.1 | ||
| NM_001080747.2 | NP_001074216.1 | |||
| NM_001080748.2 | NP_001074217.1 | |||
| NM_001080749.2 | NP_001074218.1 | |||
| NM_010365.4 | NP_034495.2 | |||
| AF017085.1 | AAC53569.1 | |||
| AF043219.1 | AAC02990.1 | |||
| AF043220.1 | AAC02991.1 | |||
| AK013348.1 | BAB28803.2 | |||
| AK145698.1 | ||||
| AK147201.1 | BAE27759.1 | |||
| AY030290.1 | AAK49785.1 | |||
| AY030291.1 | AAK49786.1 | |||
| AY030292.1 | AAK49787.1 | |||
| AY030293.1 | AAK49788.1 | |||
| BC053044.1 | AAH53044.1 |
More Information
Set Membership: |
|

Genomic Map