Assay Details
Target Gene Details
Entrez Gene ID: | 77697 |
Gene Name: | methylmalonic aciduria (cobalamin deficiency) type B homolog (human) |
Gene Aliases: |
9130222L19Rik, ATR |
Location: |
Chr.5:114431034-114444073 on Build GRCm38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mmab | NM_029956.3 | NP_084232.1 | ||
| AK020286.1 | BAB32055.1 | |||
| AK034288.1 | 8 | 683 | BAC28659.1 | |
| AK088785.1 | BAC40571.1 | |||
| AK142286.1 | BAE25010.1 | |||
| AK148421.1 | BAE28544.1 | |||
| BC022159.1 | AAH22159.1 | |||
| BC057558.1 | AAH57558.1 |
More Information
Set Membership: |
|

Genomic Map