Assay Details
Target Gene Details
Entrez Gene ID: | 108086 |
Gene Name: | ring finger protein 216 |
Gene Aliases: |
2810055G22Rik, AI647468, AU019462, C86502, F830018F18Rik, TRIAD3, UIP83, Ubce7ip1 |
Location: |
Chr.5:142990893-143113061 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnf216 | NM_080561.4 | NP_542128.2 | ||
| NM_207110.1 | NP_996993.1 | |||
| AK147861.1 | BAE28184.1 | |||
| AK149741.1 | BAE29055.1 | |||
| AK154368.1 | BAE32540.1 | |||
| AK195328.1 | ||||
| AK198180.1 | ||||
| AY572785.1 | AAS78930.1 | |||
| AY572786.1 | AAS78931.1 | |||
| BC065066.1 | AAH65066.1 | |||
| BC079540.1 | AAH79540.1 |
More Information
Set Membership: |
|

Genomic Map