Assay Details
Target Gene Details
Entrez Gene ID: | 108086 |
Gene Name: | ring finger protein 216 |
Gene Aliases: |
2810055G22Rik, AI647468, AU019462, C86502, F830018F18Rik, TRIAD3, UIP83, Ubce7ip1 |
Location: |
Chr.5:142990893-143113061 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnf216 | NM_080561.4 | 9 | 1554 | NP_542128.2 |
| NM_207110.1 | 9 | 1725 | NP_996993.1 | |
| AF361000.1 | 1 | 33 | AAK51470.1 | |
| AK089775.1 | 9 | 1515 | BAC40959.1 | |
| AK147861.1 | 9 | 1686 | BAE28184.1 | |
| AK149741.1 | 9 | 1515 | BAE29055.1 | |
| AK154368.1 | 9 | 1515 | BAE32540.1 | |
| AK195719.1 | ||||
| AK216190.1 | ||||
| AY572785.1 | 8 | 1372 | AAS78930.1 | |
| AY572786.1 | 8 | 1543 | AAS78931.1 | |
| BC065066.1 | 9 | 1528 | AAH65066.1 | |
| BC079540.1 | 6 | 650 | AAH79540.1 |
More Information
Set Membership: |
|

Genomic Map