Assay Details
Target Gene Details
Entrez Gene ID: | 75357 |
Gene Name: | RIKEN cDNA 4930557J02 gene |
Gene Aliases: |
- |
Location: |
Chr.5:34118740-34135339 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930557J02Rik | NR_040703.1 | |||
| AK016164.1 |
Target Gene Details
Entrez Gene ID: | 272158 |
Gene Name: | DNA polymerase N |
Gene Aliases: |
POL4P |
Location: |
Chr.5:34007179-34169526 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Poln | NM_001289803.1 | NP_001276732.1 | ||
| NM_001289804.1 | NP_001276733.1 | |||
| NM_181857.4 | NP_862905.1 | |||
| AK133063.1 | BAE21491.1 | |||
| AY135562.3 | AAN39837.2 | |||
| BC141101.1 | ||||
| BC145378.1 |
More Information
Set Membership: |
|

Genomic Map