Assay Details
Target Gene Details
Entrez Gene ID: | 17527 |
Gene Name: | MpV17 mitochondrial inner membrane protein |
Gene Aliases: |
Tg.Mpv17 |
Location: |
Chr.5:31140663-31154532 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mpv17 | NM_001294322.1 | NP_001281251.1 | ||
| NM_001294324.1 | NP_001281253.1 | |||
| NM_008622.6 | NP_032648.1 | |||
| AK040477.1 | ||||
| BC013452.1 | AAH13452.1 | |||
| BY088136.1 | ||||
| BY258057.1 | ||||
| BY271802.1 | ||||
| CK622051.1 | ||||
| DV062332.1 | ||||
| M36411.1 | AAA39736.1 |
More Information
Set Membership: |
|

Genomic Map