Assay Details
Target Gene Details
Entrez Gene ID: | 68770 |
Gene Name: | putative homeodomain transcription factor 2 |
Gene Aliases: |
1110054G21Rik, 9530062N20Rik, AI447096, C77923 |
Location: |
Chr.5:20758664-20882124 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phtf2 | NM_172992.3 | 10 | 1326 | NP_766580.3 |
| AK037021.1 | 10 | 1213 | BAC29673.1 | |
| AK163376.1 | 10 | 1252 | ||
| BC052841.1 | 10 | 1239 | AAH52841.1 |
More Information
Set Membership: |
|

Genomic Map