Assay Details
Target Gene Details
Entrez Gene ID: | 66506 |
Gene Name: | proteasome (prosome, macropain) assembly chaperone 3 |
Gene Aliases: |
1810042K04Rik, 4930403H09Rik, AI303239 |
Location: |
Chr.5:139823594-139827003 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Psmg3 | NM_025604.3 | 2 | 172 | NP_079880.1 |
| AK012616.1 | 2 | 173 | BAB28357.1 | |
| AK019558.2 | 1 | 562 | ||
| BC027410.1 | 2 | 85 | AAH27410.1 | |
| BC058692.1 | 2 | 158 | AAH58692.1 |
More Information
Set Membership: |
|

Genomic Map