Assay Details
Target Gene Details
Entrez Gene ID: | 66999 |
Gene Name: | mediator complex subunit 28 |
Gene Aliases: |
1500003D12Rik, AI451633, AU045690, Eg1, FKSG20, magicin |
Location: |
Chr.5:45520229-45529284 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Med28 | NM_025895.4 | NP_080171.1 | ||
| AF318060.1 | AAL26907.1 | |||
| AK003854.1 | BAB23039.1 | |||
| AK005681.1 | BAB24184.1 | |||
| AK007855.1 | BAB25307.1 | |||
| AK154249.1 | BAE32464.1 | |||
| AK201969.1 | ||||
| BC024941.1 | AAH24941.1 | |||
| BY725533.1 | ||||
| DV653139.1 |
More Information
Set Membership: |
|

Genomic Map