Assay Details
Target Gene Details
Entrez Gene ID: | 100609 |
Gene Name: | NOL1/NOP2/Sun domain family, member 5 |
Gene Aliases: |
9830109N13Rik, AI326939, Nol1r, Wbscr20, Wbscr20a |
Location: |
Chr.5:135369930-135376805 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nsun5 | NM_145414.2 | NP_663389.2 | ||
| AF412029.1 | AAM62311.1 | |||
| AK079400.1 | BAC37634.1 | |||
| AK149521.1 | BAE28935.1 | |||
| AK156651.1 | BAE33793.1 | |||
| BC051209.1 | AAH51209.1 |

Genomic Map