Assay Details
Target Gene Details
Entrez Gene ID: | 243197 |
Gene Name: | major facilitator superfamily domain containing 7A |
Gene Aliases: |
4732482E20Rik, Mfsd7 |
Location: |
Chr.5:108441052-108450260 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mfsd7a | NM_172883.2 | NP_766471.1 | ||
| AK029021.1 | BAC26248.1 | |||
| AK087226.1 | ||||
| AK171069.1 | ||||
| BC094512.1 | AAH94512.1 |
More Information
Set Membership: |
|

Genomic Map