Assay Details
Target Gene Details
Entrez Gene ID: | 76366 |
Gene Name: | mitochondrial translational initiation factor 3 |
Gene Aliases: |
2810012L14Rik, AI414549 |
Location: |
Chr.5:146951573-146963805 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mtif3 | NM_001256100.1 | NP_001243029.1 | ||
| NM_001256101.1 | NP_001243030.1 | |||
| NM_001256102.1 | NP_001243031.1 | |||
| NM_029581.4 | NP_083857.1 | |||
| AK012733.1 | BAB28438.1 | |||
| AK088146.1 | 1 | 80 | ||
| AK147090.1 | BAE27670.1 | |||
| BC052439.1 | ||||
| BC115684.1 | ||||
| BC115685.1 | ||||
| BY284518.1 |
More Information
Set Membership: |
|

Genomic Map