Assay Details
Target Gene Details
Entrez Gene ID: | 102465620 |
Gene Name: | microRNA 7025 |
Gene Aliases: |
Gm27860, mmu-mir-7025 |
Location: |
Chr.5:75176831-75176904 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mir7025 | NR_105992.1 |
Target Gene Details
Entrez Gene ID: | 18595 |
Gene Name: | platelet derived growth factor receptor, alpha polypeptide |
Gene Aliases: |
AI115593, CD140a, Pdgfr-2 |
Location: |
Chr.5:75151401-75198211 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdgfra | NM_001083316.1 | NP_001076785.1 | ||
| NM_011058.2 | NP_035188.2 | |||
| AK035501.1 | ||||
| AK081664.1 | BAC38283.1 | |||
| AK136490.1 | BAE23004.1 | |||
| AK145097.1 | ||||
| AK147267.1 | BAE27808.1 | |||
| AK163952.1 | BAE37548.1 | |||
| BC053036.1 | AAH53036.1 | |||
| M57683.1 | AAA39733.1 | |||
| M84607.1 | AAA39904.1 |
More Information
Set Membership: |
|

Genomic Map