Assay Details
Target Gene Details
Entrez Gene ID: | 231051 |
Gene Name: | lysine (K)-specific methyltransferase 2C |
Gene Aliases: |
E330008K23Rik, HALR, Mll3, mKIAA1506 |
Location: |
Chr.5:25271794-25498818 on Build GRCm38 |
Assay Gene Location: | Within Exon 54 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kmt2c | NM_001081383.1 | 54 | 14322 | NP_001074852.1 |
| AK129377.1 | 13 | 3949 | BAC98187.1 | |
| AK136029.1 | 7 | 1973 | BAE22782.1 | |
| AK148595.1 | 3 | 329 | ||
| AK161051.1 | 3 | 1067 | BAE36172.1 | |
| AK164521.1 | 3 | 598 | BAE37820.1 | |
| AY036887.1 | 4 | 1447 | AAK70214.1 | |
| AY138582.1 | 49 | 9937 | AAN11291.1 |

Genomic Map