Assay Details
Target Gene Details
Entrez Gene ID: | 231051 |
Gene Name: | lysine (K)-specific methyltransferase 2C |
Gene Aliases: |
E330008K23Rik, HALR, Mll3, mKIAA1506 |
Location: |
Chr.5:25271794-25498818 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 54 - Exon 55 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kmt2c | NM_001081383.1 | NP_001074852.1 | ||
| AK129377.1 | BAC98187.1 | |||
| AK136029.1 | BAE22782.1 | |||
| AK148595.1 | ||||
| AK161051.1 | BAE36172.1 | |||
| AK164521.1 | BAE37820.1 | |||
| AY036887.1 | AAK70214.1 | |||
| AY138582.1 | AAN11291.1 |

Genomic Map