Assay Details
Target Gene Details
Entrez Gene ID: | 58805 |
Gene Name: | MLX interacting protein-like |
Gene Aliases: |
ChREBP, WS-bHLH, Wbscr14, bHLHd14 |
Location: |
Chr.5:135089891-135138382 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mlxipl | NM_021455.4 | 6 | 757 | NP_067430.2 |
| AF156604.1 | 6 | 742 | AAF68175.1 | |
| AF245475.1 | 6 | 735 | AAK20940.1 | |
| AF245476.1 | 6 | 735 | AAK20941.1 | |
| AF245477.1 | 7 | 669 | AAK20942.1 | |
| AF245478.1 | 6 | 735 | AAK20943.1 | |
| AF245479.1 | 6 | 735 | AAK20944.1 | |
| AK143800.1 | 6 | 744 | ||
| JQ437838.1 | 6 | 603 |
More Information
Set Membership: |
|

Genomic Map