Assay Details
Target Gene Details
Entrez Gene ID: | 214642 |
Gene Name: | cadherin-like and PC-esterase domain containing 1 |
Gene Aliases: |
6720481P07, A430107O13Rik, AI552584 |
Location: |
Chr.6:21985705-22256407 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cped1 | NM_001081351.1 | 3 | 1387 | NP_001074820.1 |
| AK032959.1 | 2 | 74 | BAC28099.1 | |
| AK040587.1 | ||||
| AK087585.1 | ||||
| BC151018.1 | 3 | 902 |

Genomic Map