Assay Details
Target Gene Details
Entrez Gene ID: | 22190 |
Gene Name: | ubiquitin C |
Gene Aliases: |
2700054O04Rik, AI194771, Rps27a, TI-225, Uba52, Ubb |
Location: |
Chr.5:125385965-125390017 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ubc | NM_019639.4 | NP_062613.3 | ||
| AK004727.1 | ||||
| CF162845.1 | ||||
| CF172065.1 |

Genomic Map