Assay Details
Target Gene Details
Entrez Gene ID: | 11987 |
Gene Name: | solute carrier family 7 (cationic amino acid transporter, y+ system), member 1 |
Gene Aliases: |
4831426K01Rik, AI447493, Atrc-1, Atrc1, Cat1, Rec-1, Rev-1, mCAT-1 |
Location: |
Chr.5:148327410-148399904 on Build GRCm38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc7a1 | NM_001301424.1 | NP_001288353.1 | ||
| NM_007513.4 | NP_031539.3 | |||
| AK139342.1 | BAE23966.1 | |||
| AK147991.1 | BAE28273.1 | |||
| AK163202.1 | BAE37233.1 | |||
| BC145779.1 | ||||
| BC145781.1 | ||||
| JN226407.1 | ||||
| JN226408.1 | ||||
| JN226410.1 | ||||
| M26687.1 | AAA37574.1 |
More Information
Set Membership: |
|

Genomic Map