Assay Details
Target Gene Details
Entrez Gene ID: | 76800 |
Gene Name: | ubiquitin specific peptidase 42 |
Gene Aliases: |
2410140K03Rik, 3110031A07Rik, A630018G05Rik, D5Ertd591e |
Location: |
Chr.5:143710325-143759808 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usp42 | NM_029749.2 | NP_084025.2 | ||
| AK010801.1 | ||||
| AY336567.1 | ||||
| BC137852.1 | ||||
| BG079818.2 |
More Information
Set Membership: |
|

Genomic Map