Assay Details
Target Gene Details
Entrez Gene ID: | 231691 |
Gene Name: | serine dehydratase |
Gene Aliases: |
4432411H13Rik, SDH |
Location: |
Chr.5:120476547-120484935 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sds | NM_145565.1 | 6 | 558 | NP_663540.1 |
| AK149438.1 | 6 | 575 | BAE28875.1 | |
| BC021605.1 | 6 | 555 | AAH21605.1 | |
| BC021950.1 | 6 | 558 | AAH21950.1 |
More Information
Set Membership: |
|

Genomic Map