Assay Details
Target Gene Details
Entrez Gene ID: | 257635 |
Gene Name: | serine dehydratase-like |
Gene Aliases: |
4432411H13Rik, AI504310, SDH1, SDS-RS1, TDH |
Location: |
Chr.5:120458193-120472891 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sdsl | NM_133902.2 | NP_598663.2 | ||
| AF328927.1 | AAL56988.1 | |||
| BC022601.1 | AAH22601.1 |
More Information
Set Membership: |
|

Genomic Map